This section describes settings that should only be made on Chromeleon stations that are directly connected to instruments.
NOTE
It is not recommended to disable power management on Chromeleon client stations to which no instrument is connected.