Chromeleon license codes are backward compatible with older Chromeleon versions (7.3.0 onwards). To achieve backward compatibility of license codes with older Chromeleon versions, the License Server must always provide the latest Chromeleon version.
NOTE
Deactivate the license codes before you update to the latest Chromeleon version. Reactivate the license codes after the update has completed.