After installation of Chromeleon, the Symantec Endpoint Protection™ service may get stuck in an infinite restart loop and the message “Download insight is not functioning correctly due to an intrusion prevention component” is shown in the notification area.
To solve the problem, reinstall Symantec Endpoint Protection.