CAUTION
The following steps delete all locally stored Chromeleon data (including chromatograms, custom ePanels, instrument configuration, user database, and so on) from the PC.
  • If you want to save the data, follow the steps in the section Administering Chromeleon > Backing Up Data > Backup Procedure and Data File Location of the Chromeleon 7 Help before you continue.
Reboot the computer (or stop the SQL Server (SQLEXPRESS) service), then delete the following folder: C:\ProgramData\Dionex\Chromeleon.
NOTE
If you created local data vaults in locations other than the default path, you must delete these separately.