Chromeleon relies on several third-party software components (Installed third-party software), which are installed during the setup process. To avoid problems with software installed after Chromeleon, most of them are not removed automatically when uninstalling Chromeleon 7. You can remove several of these software components manually following steps similar to removing Chromeleon 7 (Uninstall Chromeleon 7).
CAUTION
The software components listed below might be used by various applications on your computer, not only Chromeleon 7. Removing them may corrupt those applications.
The following components are not removed by the Chromeleon 7 uninstall process:
  • Microsoft SQL Server Express
The uninstall process attempts to remove the following components:
  • Microsoft Visual C++ Redistributables