You can remove the Chromeleon program files from your computer by following the instructions below.
TIP
These steps only remove the software components. All your data and settings are preserved. This has the advantage that you can safely uninstall the software without affecting your data.
Procedure
- Insert the Chromeleon installation medium into the PC.
- Click Run Install.
- This starts the Chromeleon setup in Maintenance mode.
- Alternatively, click Control Panel > Programs and Features. Double-click the current Thermo Chromeleon program.
- This starts the Chromeleon setup in Maintenance mode.
- Click Uninstall.
- Follow the on-screen instructions to remove the program from the computer.
NOTE
Uninstalling Chromeleon does not remove the Thermo Scientific MS instrument drivers that were installed manually.