During the software installation, the standard data vault ChromeleonLocal is automatically generated (in the corresponding subfolder) in the default location.
During the installation, another local data vault (an XVault™) is created. This is a special data vault that stores a cached copy of the running sequences. The purpose of the XVault is to keep instruments running and data accessible for processing during network outages, but it is an essential part of stand-alone installations as well. The XVault is hidden.
TIP
The default standard data vault and the XVault need no configuration.
The default location for standard data vaults is:
C:\ProgramData\Dionex\Chromeleon\DataVaults\<data vault name>