The Chromeleon Administration Console provides the starting point for most administrative tasks in Chromeleon:
  • Managing licenses
  • Scheduling data transfer among data vaults, for example for archiving purposes
  • Managing Chromeleon users, groups, and privileges
  • Configuring Global Policies, for example enabling User Mode
  • Monitoring Domain Resources, for example checking which resources (instruments and data vaults) are available in a Chromeleon domain
  • Managing data vaults installed on the local computer
TIP
Some of these features are only available in a domain installation and are therefore not covered in this document. Contact your local Thermo Fisher Scientific support to learn more about the advantages a networked Chromeleon installation may bring to your laboratory.
Chromeleon Administration Console with left and right pane
Chromeleon Administration Console with left and right pane
To open the Chromeleon Administration Console select Start > All Programs > Thermo Chromeleon 7 > Administration Console.