You can start all configuration work for Chromeleon 7 from two starting points:
- From the Chromeleon Services Manager, which is a Chromeleon tool that appears minimized as the Chromeleon tray icon in the notification area of the Windows taskbar.
- From the Administration Console, which is a Chromeleon tool that can be started through the Start menu.