Load the USB driver for a module - Chromeleon CDS Software - Load the USB driver for a module - Chromeleon CDS Software - Chromeleon CDS Software
Chromeleon 7.3.2 MUa Installation Guide
- Product Tree
- Ardia Platform > Software > Chromeleon CDS Software
- Desktop Software > Chromeleon CDS Software
- Document Type
- Installation Guide
- Software version
- 7.3.2 MUa
- Turn on the computer, but not the module.
- Log in to the Windows operating system using an account with local administrator privileges.
- Turn on the module using the main power switch.
- Windows automatically detects the new module and performs the USB installation.