You can activate licenses manually using the QR code or by browsing to the web address displayed on the License Activation dialog box. If the LRC is valid and can be activated, the website will supply a LAC to enter the dialog box to complete the activation process.
To activate a license manually:
Procedure
  1. There are two ways:
  2. In the License Activation dialog box, click the hyperlink.
OR
  1. Scan the QR code using your mobile phone.
  2. The Chromeleon License Activation web portal is displayed in your default browser.
  3. Enter the License Request Code (LRC) which is displayed in the License Activation dialog box.
  4. Click Activate License.
  5. Chromeleon web portal generates the License Activation Code. Use this code to activate the Chromeleon license.
  6. In the Chromeleon application, enter the License Activation Code in the LAC text box.
  7. Click Manual Activation.
  8. The license is activated.
NOTE
The license activation status is displayed in the log area of the License activation dialog box.