Every Chromeleon installation comes with up to 60 days grace period. You need to activate the license to continue using Chromeleon after the grace period. You can use any one of the following methods to activate the license codes which are in the Pending activation, Grace Period Expired, Subscription pending activation or Subscription license expired state:
- Web activation
- Manual activation
TIP
You can navigate to the Manage Licenses dialog box by using any one of the following methods:
- Administration console > Chromeleon License Manager > Manage Licenses
- Chromeleon console > Help > About Chromeleon > License Details > Manage Licenses
To activate a license initialization code:
Procedure
- In the Manage Licenses dialog box, select the license initialization code to be activated.
- Click Activate.
- Or right-click on the selected code and click Activate.
- The license activation dialog box is displayed with an auto-generated License Request Code and a QR code.
- Click Web Activation to automatically activate the license (License web activation).
- Or click Manual Activation to manually activate the license (License manual activation).