Mass spectrometry drivers
For Chromeleon 7.3.2, mass spectrometry drivers are provided as separate installation packages which must be installed manually. For more information, see the MS Software Installation Guide provided on the Chromeleon installation medium.
USB virtual COM port
Some of the Chromeleon drivers require the installation of a USB virtual COM port. The driver software is provided as a separate installation package.
The USB Virtual COM Port folder provided in the Drivers folder of the Chromeleon installation medium contains the tested driver version. The USB Virtual COM Port folder also contains a ReadMe file with installation instructions.
Waters Driver Pack and Acquity System Driver
For Chromeleon 7.3.2, Waters Driver Pack and Acquity System Driver are provided as a separate installation package which must be installed independently by the user. For more information, documentation, and installation of the package, contact Waters Corporation.