It is not required to uninstall previous versions of Chromeleon/PeakNet 6 before installing Chromeleon 7.
The Chromeleon Setup checks current activities, for example, running queues. If a queue is running, Chromeleon reports a warning. You can select one of the following actions:
  • Ignore – Select this option if you want to ignore the warning and continue with the installation. The Instrument Controller stops, and the queue is aborted.
  • Retry – Select this option if you want to wait for the queue(s) to finish and retry the installation of Chromeleon 7.3.2. The state of the queues is checked again during the new installation attempt.
  • Cancel – Select this option to cancel the installation.