Parallel installation of different Chromeleon 7 versions (or service releases) on the same PC is not supported. When Chromeleon 7.3.2 is installed on a PC where an earlier version of Chromeleon 7 is present, the earlier version gets upgraded. For more information, see Install Chromeleon 7 – main and service release.