The chapter describes the pre-requisites, firewall settings, and the settings that should only be made on Chromeleon stations that are directly connected to instruments.
CAUTION
Data vaults become inaccessible due to change of name or network path after Chromeleon 7 installation
If you change the name or network path of the computer after the Chromeleon installation has completed, data vaults become inaccessible.
- Specify the computer name and path before you install Chromeleon.
- If you need to change the computer name after Chromeleon was installed, contact Thermo Fisher Scientific Service for support.