Refer to the document Supported Operating Systems in the Documents folder of the installation medium for information about the operating systems that are supported for use with Chromeleon 7.
NOTE
Make sure that the language of the Chromeleon package matches the language of the installed operating system. For example, install Chinese-language Chromeleon on a Chinese language operating system.
TIP
When installing Chromeleon on a Windows 10 Enterprise/Pro operating system, the option Turn on fast startup (recommended) should be disabled in the Windows 10 shutdown settings. This ensures that the Chromeleon Instrument Controller Service is stopped correctly on system shutdown and prevents potential instrument connection issues when the computer is powered up again.