Use Chromeleon to control your LCMS or GCMS - Use Chromeleon to control your LCMS or GCMS - Chromeleon 7.3.2 MS Software Installation Guide - Chromeleon CDS Software - Use Chromeleon to control your LCMS or GCMS - Chromeleon CDS Software - Chromeleon CDS Software - topic
Chromeleon 7.3.2 MS Software Installation Guide
- Product Tree
- Ardia Platform > Software > Chromeleon CDS Software
- Desktop Software > Chromeleon CDS Software
- Document Type
- Installation Guide
- Software version
- 7.3.2
- Remove the LC, GC, and MS hardware drivers from the Foundation Instrument Configuration.
- Start the Chromeleon Instrument Controller.
- Right-click the Chromeleon tray icon in the notification area of the Windows task bar.
- Click Start Chromeleon Instrument Controller.