It is possible to use Chromeleon on the same PC as Xcalibur or TraceFinder.
If you are using Xcalibur or TraceFinder and Chromeleon on the same PC, MS instrument control must be configured and connected to only one software package.
It is possible to use Chromeleon on the same PC as Xcalibur or TraceFinder.
If you are using Xcalibur or TraceFinder and Chromeleon on the same PC, MS instrument control must be configured and connected to only one software package.