With Chromeleon version 7.2.10 and higher, it is possible to install and operate Chromeleon on a PC that also has Xcalibur or TraceFinder installed.
With Chromeleon version 7.2.10 and higher, it is possible to install and operate Chromeleon on a PC that also has Xcalibur or TraceFinder installed.