To control an MS instrument using Chromeleon, the following software is required:
- A matching instrument driver for the MS instrument
NOTE
Most of the MS instrument drivers are provided as standalone installation packages that must be installed separately.
- Thermo Foundation™
For information about the Thermo Foundation version that is validated for use with the respected MS instrument driver, refer to the List of Supported Instruments. The document can be accessed through Start > Thermo Chromeleon 7 > Documents, or can be found in the Documents folder on the Chromeleon installation medium.