For controlling MS instruments in Chromeleon 7, the following licenses are required:
- Instrument Class 1 license and one of the following licenses:
- SQ/QqQ GC-MS Data Acquisition license
- - OR -
- LC-MS Data Acquisition license.
For controlling MS instruments in Chromeleon 7, the following licenses are required: