Processor | GC-MS | 2.4 GHz Dual Core Processor |
LC-MS
| 2.4 GHz Dual Core Processor | |
Recommended | 3 GHz Intel Core i7* or better | |
RAM | Recommended | 16 GB or more |
Hard disk | Single Quad systems (GCMS & LCMS) | 250 GB** |
LCMS Triple stage Quadrupole systems | 400 GB** | |
GCMS Triple stage Quadrupole systems | 250 GB | |
MSQ Plus | 150 GB | |
High Resolution Accurate Mass Systems (GCMS & LCMS) | 1 TB** | |
Optical Drive |
| DVD |
Display | Recommended | 1280 x 1024 (MSQ Plus), 1440 x 900 or higher resolution 32-bit color |
USB Ports | MSQ Plus | 1 port |
LAN Ports |
| 2 ports (Includes 1 additional LAN port for the Chromeleon Client/Server PC.) |
*A 64-bit WindowsTM version is required to take full advantage of this hardware.
**These hard disk requirements are set to enable storage of a substantial amount of data. The software itself requires approximately 4 GB disk space on the C drive (twice as much during an update). In addition, a few GB disk space must always be available on the C drive for sequences cached in the XVault.