Start the Instrument Configuration Manager - Chromeleon CDS Software - Start the Instrument Configuration Manager - Chromeleon CDS Software - Chromeleon CDS Software
Chromeleon 7.3.2 MS Software Installation Guide
- Product Tree
- Ardia Platform > Software > Chromeleon CDS Software
- Desktop Software > Chromeleon CDS Software
- Document Type
- Installation Guide
- Software version
- 7.3.2
- In the Chromeleon Services Manager, click Configure Instruments (only available if the Instrument Controller Service is running).
– OR –
- Click Start > Programs > Thermo Chromeleon 7 > Instrument Configuration Manager.