Procedure
  1. Right-click the Chromeleon Services Manager tray icon in the notification area of the Windows task bar.
  2. Click Start Chromeleon Instrument Controller.
TIP
If the Chromeleon tray icon is not visible on the task bar, click Start > Programs > Thermo Chromeleon 7 > Services Manager.