If MS software components are no longer useful, you must remove them manually from the computer.
NOTE
Both Thermo Foundation and the MS driver must be removed.
To remove the MS driver from the computer:
Prerequisites
- Prior to uninstalling, ensure that the MS is not included in the configured modules for Xcalibur.
Procedure
- Click the Start button.
- In the Windows Control Panel, click Apps > Uninstall (Windows 10) or Programs and Features (Windows 8.1).
- Find the MS driver you want to uninstall, right-click, and select Uninstall/Change.
- Follow the on-screen instructions to remove the program.
- Reboot your computer, if prompted to do so.
To remove Thermo Foundation from the computer:
Procedure
- Click the Start button.
- In the Windows Control Panel, click Apps > Uninstall (Windows 10) or Programs and Features (Windows 8.1).
- Find the entry for Thermo Foundation, right-click, and select Uninstall/Change.
- Follow the on-screen instructions to remove the program.
- Reboot your computer, if prompted to do so.