Use Chromeleon to control your LCMS or GCMS - Use Chromeleon to control your LCMS or GCMS - Chromeleon 7.2.10 MUi MS Software Installation Guide - Chromeleon CDS Software - Chromeleon 7.2.10 MUi MS Software Installation Guide - Xcalibur, TraceFinder and Chromeleon - Use Chromeleon to control your LCMS or GCMS - Installation on a PC with Xcalibur/TraceFinder installed - Chromeleon CDS Software - Chromeleon CDS Software - topic
Chromeleon 7.2.10 MUi MS Software Installation Guide
- Product Tree
- Desktop Software > Chromeleon CDS Software
- Ardia Platform > Software > Chromeleon CDS Software
- Document Type
- Installation Guide
- Software version
- 7.2.10 MUi
- Remove the LC, GC, and MS hardware drivers from the Foundation Instrument Configuration.
- Start the Chromeleon Instrument Controller.
- Right-click the Chromeleon tray icon in the notification area of the Windows task bar.
- Select Start Chromeleon Instrument Controller.