Start the Instrument Configuration Manager - Start the Instrument Configuration Manager - Chromeleon 7.2.10 MUi MS Software Installation Guide - Chromeleon CDS Software - Chromeleon 7.2.10 MUi MS Software Installation Guide - Start the Instrument Configuration Manager - Install and configure MS software components - Configure MS instruments in Chromeleon - Chromeleon CDS Software - Chromeleon CDS Software - topic
Chromeleon 7.2.10 MUi MS Software Installation Guide
- Product Tree
- Ardia Platform > Software > Chromeleon CDS Software
- Desktop Software > Chromeleon CDS Software
- Document Type
- Installation Guide
- Software version
- 7.2.10 MUi
- In the Chromeleon Services Manager, select Configure Instruments (only available if the Instrument Controller Service is running).
– OR –
- Select Start > Programs > Thermo Chromeleon 7 > Instrument Configuration Manager.