NOTE
The upgrade from BioPharma Finder 3.2 (or earlier) to BioPharma Finder 5.4 is not directly supported. You must perform a stepwise upgrade first to BioPharma Finder 4.0 QF1, and then to BioPharma Finder 5.4.
If you have BioPharma Finder 3.2 or prior and attempt to install BioPharma Finder 5.4, the installer will display a warning that the upgrade is not supported.
See the Installation Instructions for more information.
During the upgrade of the BioPharma Finder software, two main folders will automatically be backed up:
  1. C:\ProgramData\ThermoScientific\Databases
  2. C:\ProgramData\ThermoScientific\BioPharma
Copies of these files will be placed in C:\ProgramData\ThermoScientific\BioPharma and named using the following notation: <<legacy application version number>>_backup_DateAndTime.
NOTE
When the software has been successfully installed and upgraded, the backup files can be deleted or moved to another archive location to save space on the processing computer.