Follow these instructions to install the BioPharma Finder software, version 5.4. You must be a system administrator on the installation computer to install the software. You may license the application as either an administrator or a standard user.
Install the BioPharma Finder 5.4 software
Procedure
- Close all open Thermo Scientific applications.
- Do one of the following:
- Download and unzip the
BioPharma Finder 5.4 − Installer.zipfile from the Flexera™ Thermo Scientific Software Download and Licensing Portal account. Then, double-clickBioPharmaFinderSetup.exeto launch the installer wizard. - Insert the BioPharma Finder 5.4 USB flash drive into the computer.
- The Autorun program automatically launches the installer wizard.
- If the installer wizard does not start, navigate to the USB drive location in Windows Explorer and double-click
BioPharmaFinderSetup.exe. - In the installer wizard, do the following:
- Select the License Agreement tab to view the terms and conditions of the end-user license agreement and then select the I agree to the license terms and conditions checkbox.
- Select the System Requirements tab to check the system requirements.
- Select the Documents tab to view the Installation Instructions, Release Notes, User Guides, Example Data, and other documents for the BioPharma Finder 5.4 software.
- When you are finished reviewing the tabs, select Install (for new software users) or Upgrade (for upgrading software users) to proceed with the installation process.
- If the installer detects previous versions of the BioPharma Finder software on your system, select Yes at the prompt to uninstall all previous software versions and upgrade your legacy data.
- The installer wizard automatically backs up your legacy results onto the computer and upgrades these results to be compatible with the new software version.
- Once installation is complete, on the installer Welcome page, select IQ Report to review the IQ report (found in
C:\Program Files\Thermo\IQ Reports), select Launch to open the software and activate a license, or select Close to exit the installer wizard.