Use the Flexera Thermo Scientific Product Licensing wizard to activate (or deactivate) the BioPharma Finder application. This wizard offers several different licensing options: all workflows, or individual Peptide Mapping Analysis, Oligonucleotide Analysis, Intact Mass Analysis, and Top Down Analysis, workflows as well. Activation of these features depends on the product purchased and is controlled by the license activation key. If you would like to add feature to the software, contact your customer service representative.
Before you transfer the license to another computer, you must deactivate the existing BioPharma Finder application and then transfer the license key. If you upgrade from previous versions of BioPharma Finder, Thermo Scientific PepFinder™, or Thermo Scientific Protein Deconvolution™, you must obtain a new license key for the BioPharma Finder application.
Activate (or deactivate) a BioPharma Finder 5.4 license
Procedure
  1. Open the License Activation wizard as follows:
  2. Open the BioPharma Finder application.
  3. Select Help > About BioPharma Finder to display the License Activation wizard.
  4. Select the Activate (or Deactivate) button to start the activation or deactivation process.
  5. To activate the demo version of the software, select the 60-day trial license.
  6. For purchasers of the software, set a permanent license using the activation code as follows:
  7. Log in to your Flexera Thermo Scientific Software Download and Licensing Portal account.
  8. In the left navigation pane, under Software & Services, select Order History.
  9. From the list of ordered products, select the order number. The Order Details table provides the activation code in the last column. The order number is in the Thermo Scientific “Your Order is Ready” email message.
  10. Continue through the License Activation wizard to activate the license using the activation code.
NOTE
You cannot use an activation code from an earlier version of the BioPharma Finder software to activate BioPharma Finder 5.4.