Suggested recovery actions
  • For some issues, restarting the application is the appropriate recovery action.
  • As a fix, we generally do not recommend reinstalling the software or the operating system, which more commonly occurs after you install a new hard drive.
  • After the installation on the software on some PCs, there is an issue with viewing the IQ report using the displayed link. If this occurs, the IQ report can be found in C:\Program Files\Thermo\IQ Reports.
Feature requests and other omitted items
  • We do not include issues where there is insufficient information logged to successfully reproduce the reported problem.
  • We do not list feature requests as software issues, regardless of the reported significance or severity of the request. Product managers evaluate logged feature requests for future releases.
  • We report only discrepancies in the documented software as known issues.
Terminology
The following tables define the terms used to describe the severity and risks associated with the known defects detailed in the Known Defects for BioPharma Finder 5.4 table. Product management assesses risk, which can differ significantly from the reported severity.
Table Defect Severity
Severity
Interpretation
Critical
A problem that renders the system unusable because either an entire function is unusable and no workaround exists or use of the current system compromises data integrity or results in data loss. Catastrophic problems also include significant and non-obvious quantitative errors and all human and instrument safety issues.
High
A serious issue that does not affect data integrity (meaning data loss, corruption of data, or the wrong answer), but affects the customer’s ability to use the product as designed. It can be a failure, design issue, or documentation error or omission. A workaround might or might not exist.
Medium
A minor error or poor behavior of a product feature. There is likely a workaround.
Low
An issue that has a limited effect on customer usage of the product, including defects with visibility so low that a customer might never see it or ease-of use-issues or other items not causing any performance degradation.
Table Defect Risk
Risk
Interpretation
High
Occurrence is likely to happen and can compromise operation.
Medium
Occurrence is uncommon, but if it occurs, can compromise operation.
Low
Issue is minor; however, the software might operate differently from a user’s expectations. A workaround is often available.
No risk
This issue causes no problems but is a common inconsistency or an issue with a low severity level.
Known defects
The following table lists the known defects in the BioPharma Finder 5.4 software, categorized by software section, including a brief description and information related to each defect’s severity and risk. The item ID is the internal number assigned to each issue.
Table Known defects in BioPharma Finder
Severity
Risk
Item ID
Software area
Description
High
High
(TFS) 9716
General
Images copied from the BioPharma Finder application and pasted into Microsoft Word or PowerPoint™ that are then saved as PDF images might be corrupted.
High
High
(TFS) 32986
Intact Mass Analysis
Duplicate copies of the chromatogram and deconvoluted and source spectra (from the same raw file) are created for Multiconsensus experiments when the raw data file contains more than one component that satisfies the multiconsensus merge parameters.
High
Medium
40410
Intact Mass Analysis
Running an experiment using the “Auto ReSpect” processing method does not identify the expected components.
High
Low
38374
Installation
When upgrading from BioPharma Finder version 3.0 to 3.1, if there is not enough disk space and the user selects “Ignore” in the warning message, the application still attempts to create a backup folder and the installation fails.
Workaround: First, uninstall version 3.0 and then install version 3.1 as a clean install.
High
Low
38699
39292
Peptide Mapping Analysis
The “ID Type” value (Full, MS2) displayed for some components in the Processing and Review > Results table is incorrect.
Workaround: Select the component in the Results table and then select the MS2 Spectra tab to check if a MS2 spectrum is displayed for the component.
Medium
Medium
(TFS) 81000
Installation
When the BioPharma Finder installer is run on some Windows 7 machines, the installer stalls at the "Initializing" step.
Medium
Medium
49511
Intact Mass Analysis
When using the “Default Native Above 1 Million” processing method for QB files, users might not be able to set an appropriate retention time (RT) range.
Medium
Medium
(TFS) 14462
Intact Mass Analysis
When a user is connected to a remote desktop computer and runs the BioPharma Finder application on a trial license, the Intact Mass Analysis workflow is disabled.
Medium
Medium
51171
Intact Mass Analysis
Top Down Analysis
On the Parameters > Component Detection page of the processing method editing wizard, the “Resolution at 400 MHz” parameter is incorrect.
Medium
Medium
185972
Oligonucleotide Analysis
The theoretical masses of some modifications in the Oligonucleotide Analysis workflow are incorrectly calculated for full-scan MS data.
Medium
Medium
179638
Peptide Mapping Analysis
In the Theoretical Protein/Peptide Manager, custom proteases that cleave at both the C- and N-terminals might not cleave correctly at the N-terminal.
Medium
Medium
39357
Peptide Mapping Analysis
Intact Mass Analysis
Users are able to save a processing method that contains errors.
Medium
Medium
52372
Peptide Mapping Analysis
Oligonucleotide Analysis
In the Process and Review > Results table, the “Peptide ID” values include component mass (m) information, but the “Mono. Mass Exp.” values are “0.000”.
Medium
Medium
747861
Reporting
After navigating pages containing charts, the Report utility produces an error when the user selects the “Preview” button.
Medium
Low
749147
(INC) 758078
Ardia Platform
There is a registration error when upgrading from BioPharma Finder version 5.1 to version 5.2 or 5.3 when connected to the Ardia Platform version 1.1.
Workaround: If you encounter this error, please contact Technical Support at digital.support@thermofisher.com for assistance.
Medium
Low
758151
Ardia Platform
When a user updates the raw data file paths associated with an experiment on a given PC, on a second PC, the experiment either cannot be opened from the Ardia Platform data repository or the application prompts the user to specify the raw data files.
Workaround: After updating the raw data file paths, on the original PC, save the experiment in a new location in the Ardia Platform data repository before opening the experiment on the second PC.
Medium
Low
123001
Installation
If a user declines the EULA when installing BioPharma Finder version 3.0 (or earlier), subsequently attempting to upgrade to version 4.0 (or later) fails.
Medium
Low
119656
Installation
Post-installation IQ Report link does not work.
Workaround: Access the IQ Report via C:\Program Files\Thermo\IQ Reports.
Medium
Low
52888
Intact Mass Analysis
For experiments using the sliding windows deconvolution method with a single raw data file, the “Matched Mass Error % CV” value should be “0.”
Medium
Low
39296
Intact Mass Analysis
After recalculating the average DAR using selected components, the drug loads are zero for non-selected components.
Workaround: Uncheck all components and select “Recalculate”.
Medium
Low
39318
Intact Mass Analysis
When reprocessing an experiment with updated sequence matching/DAR parameters, the application should not reprocess the entire deconvolution experiment, but only the updated tasks.
Medium
Low
39279
Intact Mass Analysis
The retention time (RT) range displayed on the chromatogram is incorrect (smaller) than the range specified in the raw data file.
Medium
Low
122165
Peptide Mapping Analysis
When BioPharma Finder version 3.0 (or earlier) is upgraded to version 4.0 (or later), the Beginning Peak Width and Ending Peak Width parameters display “0” for Peptide Mapping Analysis experiments.
Workaround: After upgrading, use the real-time optimization (RTO) feature to set the Beginning Peak Width and Ending Peak Width parameters to the same value as the Typical Chromatographic Width parameter. Save the method and reprocess the experiment.
Medium
Low
178056
Peptide Mapping Analysis
For host cell protein (HCP) experiments, the default “Find All Ions with MS/MS” task may generate components with associated MS2 spectra that have both average and monoisotopic masses of “0”.
Medium
Low
164107
Peptide Mapping Analysis
For host cell protein (HCP) experiments, HCP identifications are incorrect for protein sequences with a negative variable terminal modification.
Medium
Low
177288
Top Down Analysis
The application may freeze when the user is loading or viewing experiment results with more than 25 proteoforms.
Workaround: Reduce the total number of proteoforms searched in per experiment by performing multiple experiments with less than 10 proteoforms per experiment.
Low
Low
466619
(INC)758085
Ardia Platform
When signing out of the Ardia Platform via ThermoAD IdP, the user is not automatically signed out of the current BioPharma Finder session.
Low
Low
38718
General
The application does not fully support display on 4K monitors.
Low
Low
647675
Installation
When downgrading from version 5.0 (or later) to 3.0 (or earlier), the expected warning message is not displayed.
Low
Low
56143
Intact Mass Analysis
For an experiment containing multiple raw data files, the deconvoluted spectrum is not displayed due to a refresh error after filtering component results.
Low
Low
51273
Intact Mass Analysis
When the run queue is empty, the Queue page table header is missing.
Low
Low
39295
Intact Mass Analysis
When processing an experiment in Manual mode, specifying a retention time (RT) range greater than the range of the actual chromatogram causes the chromatogram and source spectrum to be empty.
Low
Low
39283
Intact Mass Analysis
Single-scan QB raw data files cause the source spectrum to be blank when switching from Sliding Windows to Average Over Selected RT deconvolution.
Workaround: Create a processing method for a Sliding Windows deconvolution with a “Target Avg. Spectrum Width” parameter set to less than 0.02(RT).
Low
Low
40032
Intact Mass Analysis
On the Parameters > Deconvolution Parameters (ReSpect) page of the processing method editing wizard, the “Resolution at 400 MHz” parameter is incorrect for experiment containing both FTMS and ITMS full-scan data.
Low
Low
40011
Intact Mass Analysis
When there are many jobs submitted to the run queue, experiment processing is stalled for an experiment using the Xtract with Sliding Windows deconvolution method.
Workaround: Delete the stalled experiment from the run queue and then resubmit the experiment once the run queue is smaller.
Low
Low
39544
Intact Mass Analysis
When the retention time (RT) range is manually set to [0.0 to 0.0] for a Sliding Windows experiment, the expected warning message is not displayed when the experiment is subsequently submitted for processing in Automatic mode.
Workaround: Set the RT range to an appropriate value.
Low
Low
39358
Intact Mass Analysis
Drug load cannot be set for a component with no identification.
Low
Low
39350
Intact Mass Analysis
On the Parameters > Identification page of the processing method editing wizard, the list of modifications in the “Select a variable modification candidate for the DAR calculation” area is missing several N- and C-terminal modifications.
Low
Low
39325
Intact Mass Analysis
When other components are selected in the Process and Review > Average DAR table, users cannot update the drug load value for an unchecked component.
Workaround: Select the component in the Results table before updating the drug load value.
Low
Low
39322
Intact Mass Analysis
If a raw data file does not contain any matches for a DAR experiment, the “Raw File Name” column in the Process and Review > Average DAR table is blank.
Low
Low
39304
Intact Mass Analysis
For a ReSpect with Sliding Windows deconvolution experiment, the “Mass Std Dev” and “PPM Std Dev” parameters are shown in the deconvolution report, but not in the Results table or exported results file.
Low
Low
39291
Intact Mass Analysis
When a ReSpect deconvolution experiment is submitted to the run queue, the experiment is processed, but the “Number of Components Detected” column displays “0” if the raw file name, experiment name, or file path exceed 256 characters.
Workaround: Keep file names, experiment names, and file paths under 256 characters.
Low
Low
39288
Intact Mass Analysis
For a Multiconsensus experiment with raw data files containing multiple peaks, only the first component is exported for each peak when a user exports the top-level results as a Microsoft Excel™ file.
Workaround: Export all components as an Excel file.
Low
Low
38720
Intact Mass Analysis
Enhancement request: Optimize layout of elements on the Process and Review page.
Low
Low
38717
Intact Mass Analysis
For an Automatic Peak Detection experiment, the extracted ion chromatogram (XIC) might not appear on the Process and Review page.
Workaround: Reload the experiment results.
Low
Low
38704
Intact Mass Analysis
If the run queue is paused and BioPharma Finder is upgraded to version 3.0, a dialog box appears asking the user if they would like to restart the run queue. If the user selects “Yes”, the application crashes.
Workaround: Process all experiments in the run queue before upgrading BioPharma Finder to a later version.
Low
Low
39374
Intact Mass Analysis
If experiment data is collected at multiple resolutions, to obtain best results, the user must run multiple experiments in which each processing method specifies a particular resolution value.
Low
Low
40211
Intact Mass Analysis
On the Parameters > Chromatogram page of the processing method editing wizard, the user can specify different retention time (RT) ranges in the “Chromatogram Parameters” and “Sliding Windows Parameters” areas.
Low
Low
51905
Licensing
In the License Activation dialog box, if a user declines to activate a permanent license and then selects “Back” to activate a trial license instead, the application gives an error.
Workaround: Close and restart the application. In the License Activation dialog box, activate a trial license.
Low
Low
38713
Licensing
During an offline deactivation, the response file for "Deactivation.req" is incorrectly named "Activation.xml".
Low
Low
117056
Oligonucleotide Analysis
Oligonucleotide sequences with a 3’-terminal modification are not completely highlighted in the Process and Review > Real Time Optimization > Oligo Sequence Map area.
Low
Low
117286
Oligonucleotide Analysis
Oligonucleotide Analysis only supports one variable modification for a given oligonucleotide sequence.
Low
Low
117636
Oligonucleotide Analysis
When users export checked components in the Modification Summary > Components table “As Displayed”, the file is blank when viewed with Apache™ OpenOffice™.
Workaround: Export all or checked components in the Modification Summary table as an Excel worksheet.
Low
Low
117583
Oligonucleotide Sequence Editor
An error results when users import FASTA files containing oligonucleotide sequences in two-letter code format.
Workaround: Import FASTA files containing sequences in single-letter or triplet code format.
Low
Low
38703
Peptide Mapping Analysis
Loading an incorrectly formatted MSQC file causes an unexpected dialog box to appear.
Low
Low
38709
Peptide Mapping Analysis
After submitting an experiment to the run queue, the Peptide Mapping Analysis page shows the last used processing method checked.
Low
Low
54723
Peptide Mapping Analysis
The application displays an unexpected arrangement of workflow page tabs following experiment processing.
Low
Low
53410
Peptide Mapping Analysis
Users might experience a refresh error on the Mapping > Modification Summary page after performing manual integration of the chromatogram.
Low
Low
39281
Peptide Mapping Analysis
When an experiment is deleted from the Load Results page, the Mapping tab is not removed.
Workaround: Close and restart the application.
Low
Low
40198
Peptide Mapping Analysis
The ∆ ppm value for a component with a “Gasphase-NH3Loss” modification is larger than expected.
Low
Low
40110
Peptide Mapping Analysis
When a user subsequently loads a new raw data file, it overwrites the previously added file.
Low
Low
40081
Peptide Mapping Analysis
When there is no protein sequence coverage determined for an experiment, the signal intensity color coding is incorrect.
Low
Low
39528
Peptide Mapping Analysis
For dimers, the MS2 spectrum shows the predicted data for a single peptide instead of the dimer.
Workaround: Confirm the presence of the dimer using the full-scan MS data; MS2 data displays the fragmentation of the constituent peptide monomers.
Low
Low
39527
Peptide Mapping Analysis
When a user performs a disulfide bond search, the ∆ ppm value is larger than expected for components predicted to be Na+ or K+ adducts.
Low
Low
38701
Peptide Mapping Analysis
The application selects the activation type as ETD over higher-quality CID in the MS2 spectrum.
Workaround: Right-click on the MS2 spectrum, select “Predict Peptide MS/MS (Kinetic Model)", and then select the correct fragmentation method in the dialog box.
Low
Low
38389
Peptide Mapping Analysis
In the Target Peptide Workbook Editor, if you edit the retention time (RT) range of a peptide such that it creates an error, the red error message on the communicator bar does not disappear even after fixing the RT range.
Workaround: Select the row for another peptide in the workbook to clear the error message.
Low
Low
38387
Peptide Mapping Analysis
For a very large experiment (100 raw data files), the Process and Review page > Trend MS Area is missing the raw data file labels on the x-axis due to space limitations.
Low
Low
38384
Peptide Mapping Analysis
In the Process and Review > Results table, resetting the filter for an individual column does not return the column to its default state.
Low
Low
38378
Peptide Mapping Analysis
For targeted experiments, the “% Abundance” values displayed on the Mapping > Modification Summary page might be incorrect for peptides with “nonunique” modifications
Low
Low
38377
Peptide Mapping Analysis
Multiple labels for a single glycan might appear in the MS2 spectrum.
Low
Low
38376
Peptide Mapping Analysis
In the Process and Review > Results table, the “Mono Mass Exp.” value for some components might differ slightly between a targeted and non-targeted experiment.
Low
Low
39302
Peptide Mapping Analysis, Intact Mass Analysis
When a user is connected to a remote desktop (Windows 10 machine) and attempts to open or load experiment results, the application displays a blank screen instead of opening the Process and Review page.
Workaround: Close and restart the application, load or open the experiment results again. The results are displayed on the Process and Review page.
Low
Low
38719
Protein Sequence Editor
When a user pastes peptide chains beginning with “> name” into the Sequence Box, the Protein Sequence Manager does not correctly differentiate the chains.
Low
Low
645518
Reporting
When switching between modules in the Reporting tool, the user is prompted to save the report template before saving the report, even if no changes to the template have been made.
Low
Low
750629
Run queue
Canceling an experiment in a given workflow with a “Preparing” status in the run queue also cancels other experiments with the same name in other workflows.
Low
Low
38738
Top Down Analysis
On the Parameters > Identification page of the processing method editing wizard, UI items are not properly aligned.
Low
Low
38712
Top Down Analysis
After inputting experiment information on the Top Down Analysis page, selecting the Home tab triggers a warning message that the application will clear the experiment settings.
Low
Low
38700
Top Down Analysis
On the Parameters > Component Detection page of the processing method editing wizard, if the “Intact Deconvolution” parameter is unselected, the m/z range displayed on the source spectrum header does not correspond to the range set in the processing method parameters.
Workaround: In the Intact Deconvolution area on the Parameters > Component Detection page of the processing method edition wizard, manually change the “m/z range” parameter and save the method.
Low
Low
38698
Top Down Analysis
On the Top Down Analysis page, if you select a protein sequence for the experiment, but then delete the sequence from the Global Reference table on the Parameters > Identification page of the processing method editing wizard, proteoforms associated with the deleted sequence are not included in the experiment results.
Workaround: Do not delete sequences from the Global Reference table if the sequence is used in an experiment that is currently processing or submitted to the run queue.