The following table lists the minimum and recommended hardware and software configurations required for BioPharma Finder 5.4 operation.
System
Minimum Requirements
Recommended Requirements
Hardware
  • Memory: 32 GB RAM
  • CPU: 8 cores
  • Storage: 100 GB
  • Display resolution: 1920 × 1080 (SXGA)
  • Optional: DVD/CD-ROM drive
  • Memory: 64 GB RAM
  • CPU: 8 cores or higher
  • Storage: 2 TB solid-state hard drive (for best performance)
  • Display resolution: 1920 × 1080
  • Optional: DVD/CD-ROM drive
Software
  • Microsoft® Windows™ 10 (English) (64-bit)
  • Microsoft® .NET Framework™ 4.8
  • Microsoft® Office 365™
  • Adobe® Acrobat® Reader DC™
  • Microsoft® Windows™ 11 (English) (64-bit)
  • Microsoft® .NET Framework™ 4.8
  • Microsoft® Office 365™
  • Adobe® Acrobat® Pro DC™
IMPORTANT
BioPharma Finder 5.4 is not compatible with Ardia Platform 1.0. You must upgrade to Ardia Platform 1.1 to connect BioPharma Finder 5.4 to the Ardia Platform.
To upgrade to Ardia Platform 1.1, contact Technical Support at digital.support@thermofisher.com.