The BioPharma Finder application matches detected components within a mass tolerance to user-defined sequences. The sequences are first categorized by sample type (protein or oligonucleotide), followed by category (experiment type), and, for peptide mapping experiments, finally by specific workflow (targeted or hydrogen-deuterium exchange, if applicable).
Adding a sequence is optional for most of the experiment workflows, except for top down analysis.