You can remove jobs in the run queue by using the Queue Manipulation button on the command bar to remove selected, completed, or all jobs.
Remove selected jobs from the run queue
Procedure
- Select the Queue tab to open the Queue page.
- In the table, select the job or jobs to remove from the run queue.
TIP
You can select multiple jobs simultaneously by selecting contiguous job names pressing the SHIFT key or noncontiguous job names pressing the CTRL key.
- On the command bar, select Queue Manipulation and then select Remove Selected.
- When prompted, select Yes in the dialog box to proceed.
NOTE
You cannot remove a job when it is still processing and its status is Running.
- The selected jobs are removed from the run queue on the Queue page.
Remove completed jobs from the run queue
Procedure
- Select the Queue tab to open the Queue page.
- On the command bar, select Queue Manipulation and then select Remove Completed.
- When prompted, select Yes in the dialog box to proceed.
- All jobs with a Completed status are removed from the run queue on the Queue page.
Remove all jobs from the run queue
Procedure
- Select the Queue tab to open the Queue page.
- On the command bar, select Queue Manipulation and then select Remove All.
- When prompted, select Yes in the dialog box to proceed.
NOTE
You cannot remove a job when it is still processing and its status is Running.
- All jobs are removed from the run queue on the Queue page.