Add custom modifications - Add custom modifications - BioPharma Finder 5.2 Top Down Analysis User Guide - BioPharma Finder Software - Add custom modifications - BioPharma Finder Software - BioPharma Finder Software - topic
BioPharma Finder 5.2 Top Down Analysis User Guide
- Product Tree
- Ardia Platform > Software > BioPharma Finder Software
- Desktop Software > BioPharma Finder Software
- Document Type
- User Guide
- Software version
- 5.2
- In the Protein Sequence Editor, select the title bar for the Modification Editor.
- The Modification Editor pane opens, displaying all of the N-terminal, C-terminal, and side chain modifications available.
- Select Add in the section corresponding to the type of custom modification you want to create (N-terminal, C-terminal, or side chain).
- In the Add New Modification dialog box, enter the appropriate information for your custom modification, including a unique modification name, monoisotopic mass, average mass, and chemical formula (optional). For custom side chain modifications, you can also specify the residues for the modification.
- See Modification Editor pane parameters.
- Select OK.
- The application saves the custom modification and the modification appears in purple in the modification list.
- You can add your custom modification to the default list of modifications for quick loading.
- See Change the default modifications for protein sequences.