Within the default list of modifications, the BioPharma Finder application sets the modifications shown in the following table as a default sublist that you can quickly load to assign as C-terminal or side chain modifications to a protein sequence.
N-terminal modifications: |
(None) |
C-terminal modifications: |
Lys |
Side chain modifications: |
Deamidation (N) |
Double oxidation |
Glycation |
H2O loss |
Hydroxylation |
Mannosylation (S) |
NH3 loss |
Oxidation (MW) |
Isomerization |