The following table describes the information in the Target Match Sequence table at the bottom of the Intact Deconvolution Matched Sequence pane.
Target Match Sequence table columns
Column
Description
(Check box)
Select this checkbox to view the sequence match identification in a particular table row.
You can select only one identification row in the table at a time.
When you select a different identification, the Protein Name and Modification columns in the Intact Deconvolution Results table automatically update based on your selection.
Protein Name
Modification
Matched Mass Error (ppm/Da) or Matched Mass Error (ppm/Da) (mean)
These columns vary depending on the number of loaded raw data files and the results format (Batch Processing or Multiconsensus) used for the experiment.
See "Additional Results table parameters for target sequence matching experiments" in the Intact Mass Analysis User Guide for more information on these columns.