You can select a different identification for the selected component in the Intact Deconvolution Matched Sequence pane on the Process and Review page.
Prerequisites
- You have loaded the experiment results and are viewing the Intact Deconvolution Matched Sequence pane on the Process and Review page.
Procedure
- On the Process and Review page, in the Intact Deconvolution Results table, select the row of a matched component.
- In the Target Match Sequence table of the Intact Deconvolution Matched Sequence pane, select the checkbox for a specific identification row.
- See Target Match Sequence table parameters.
- The new selection overwrites the default best matched sequence identification.
- Selecting a new identification automatically updates the identification results for the component in the Intact Deconvolution Results table and the corresponding peak label in the Intact Deconvolution Deconvoluted Spectrum on the Process and Review page.
- (Optional) After you make modifications to the identifications, select Save Results As in the command bar of the Process and Review page to save to a new experiment or overwrite the current experiment results.