Right-clicking the table in the ProSightBP Output pane on the Process and Review page opens a shortcut menu that contains the commands listed in the following table.
Command | Description |
|---|---|
Export All | Exports all of the results in the table as a Microsoft™ Excel™ worksheet (.xlsx). |
Export Selected | Exports only the selected results in the table as a Microsoft™ Excel™ worksheet (.xlsx). |
Component Level Only | Exports the results only at the fragment ion (top) level in the table. |
Component and Charge State Levels | Export the results at all levels in the table. |