The following table describes the information in the Matching Fragment Detail table.
This table appears when you double-click the colored bar or bars on the map in the ProSightBP Fragment Map pane, indicating one or more identified fragment ions.
Column | Description |
---|---|
Name | Displays the name of the identified fragment ion. |
Theoretical Mass | Displays the theoretical mass of the fragment ion, measured in Da. |
Observed Mass | Displays the observed mass of the fragment ion, measured in Da. |
Mass Difference (Da) | Displays the difference between the observed and theoretical fragment masses, measured in Da. |
Mass Difference (ppm) | Displays the difference between the observed and theoretical fragment masses, measured in ppm. |