The following table describes the Process command on the Process and Review page.
Command | Description |
|---|---|
Process | Reprocesses the top down analysis experiment and deconvolves the source spectra with any changes made in the Real-Time Optimization pane on the Process and Review page. To activate the Process button, you must modify the experiment parameters. See Use real-time optimization (RTO) to reprocess results. Select the Process button to open a dialog box where you can enter a new experiment/method name or retain the same name to overwrite previously saved results/parameters in the current experiment with the new data. Use only alphanumeric, space, underscore "_", and period "." characters, up to 50 maximum, in the experiment and method names. You cannot overwrite an experiment that is currently submitted to the run queue or is actively processing or canceling. You also cannot overwrite a default method. |