The following table describes the information in the Sequences Added to Experiment and Global Sequence Reference tables on the right side of the Identification page.
Identification page tables
Column
Description
Sequences Added to Experiment table
Displays the sequences added to the experiment after you select Add to Experiment in the Global Sequence Reference table.
Buttons
Remove
Removes the sequence from the Sequences Added to Experiment table.
Show Details
Displays the full protein sequence.
Global Sequence Reference table
Lists all of the existing protein sequences for top down analysis.
The columns in this table are the same as those on the Sequence Manager page.
Buttons
New
Opens the Protein Sequence Editor so that you can import a FASTA file or manually create a new protein sequence.
Edit
Opens the Protein Sequence Editor so that you can edit the selected sequence.
Delete
Deletes the selected sequence from the Global Sequence Reference table.
Add to Experiment
Adds the selected sequence to the experiment.
The added sequence appears in the Sequences Added to Experiment table.