Right-clicking inside the Chromatogram, Peak # − Intact Fragmentation Source Spectrum, or Peak # − Intact Deconvolution Source Spectrum panes of the Component Detection page opens a shortcut menu with the commands listed in the following table.
Commands | Description |
|---|---|
Reset Scale | Restores the pane to the original view. |
Copy as Displayed | Copies the image viewable in the pane to the clipboard. |
Copy per Global Settings | Copies the image in the pane to the clipboard based on the global image dimensions set in the Top Down Analysis Settings dialog box. |
Zoom Out | Shrinks the view in the pane by a factor of 2. |
Zoom In | Enlarges the view in the pane by a factor of 2. |
Chromatogram Trace Type | (For the Chromatogram pane only) Selects which type of chromatogram to display in the Chromatogram pane (TIC or BPC). |