The BioPharma Finder application provides the Top Down Default Method processing method for top down analysis experiments.
When you select Start Processing on the Top Down Analysis page, the processing method editing wizard opens and prompts you to edit the component identification and identification parameters in the method and save them to a new file to create a custom method for your experiment.
NOTE
For top down analysis experiments, you must run an experiment to edit the processing method.
You cannot edit a method without actively running an experiment.