The BioPharma Finder application includes some features from the Pinpoint™ application for managing targeted proteins and peptides.
These features provide a direct connectivity between the BioPharma Finder application and the Thermo Scientific™ Chromeleon™ chromatography data system for targeted monitoring of peptides or critical quality attributes. In addition, you can save all or selected peptides to a workbook to use as a protein sequence for a targeted peptide mapping experiment.
These features help you perform the following tasks:
  • Enter targeted proteins and proteotypic peptides.
  • For targeted proteins, perform digestion and predict the theoretical peptide fragments.
  • Create a mass list of the peptides (unmodified native peptides and modified peptides) using high resolution accurate mass (HRAM) transitions for processing.
  • Modify any of the amino acids in the protein or peptide sequence.
  • Export a targeted mass list (peptide workbook) to file format compatible with the Thermo Scientific™ Chromeleon™ chromatography data system.