The following table describes the data in the All Possible Proteoforms table.
Column | Description |
|---|---|
Row number (bolded) | The number assigned to each visible proteoform row in the table. This sequential numbering does not change when you sort or filter the table. |
Check box | Select this checkbox if you want to save the proteoforms in the selected rows with the protein sequence for a search. To select or clear all of the checkboxes at once, select or clear the checkbox in the column header. If you filter the table, the following occurs:
|
Identification | Displays the protein sequence name, followed by an underscore, and then site(modification) for each modification separated by a comma. For example, in the identification of "Protein_C4(Oxidation),N35(Deamidation)":
|
Modifications | Displays the modifications listed in the Identification column. |
Sites | Displays the sites listed in the Identification column. |
Num of Mods | Displays the number of modifications occurring in the proteoform. This number is within the range of the Number of Modifications per Proteoform values. |
Mono. Mass | Displays the monoisotopic mass of the proteoform. |
Avg. Mass | Displays the average mass of the proteoform. |