Save a protein or peptide sequence under a new name - BioPharma Finder Software - Save a protein or peptide sequence under a new name - BioPharma Finder Software - BioPharma Finder Software
BioPharma Finder 5.2 Top Down Analysis User Guide
Product Tree
Ardia Platform > Software > BioPharma Finder Software
Desktop Software > BioPharma Finder Software
Document Type
User Guide
Software version
5.2
In the Protein Sequence Editor, select Save As New in the command bar. The Save As New dialog box opens. Save As New dialog box In the New Sequence Name box, type the new name of the protein sequence. (Optional) In the Description box, type a description of the protein sequence. Select OK . Information from the new saved sequence populates the columns of the sequence table on the Sequence Manager page.