The following table lists the N-linked glycans, sorted by human host cell-line type, that are included in the N-glycan-specific search.
In the table header, "Mono mass" refers to monoisotopic mass, "Man" refers to mannose, "Fuc" refers to fucose, and "Bis Gn" refers to bisecting glycoprotein.
# | Glycan name | Antenna | Formula | Mono mass | Man | Fuc | Bis Gn | |||
|---|---|---|---|---|---|---|---|---|---|---|
1 | 2 | 3 | 4 | |||||||
1 | M3 |
|
|
|
| C34H56N2O25 | 892.317 | 3 | 0 | 0 |
2 | M4 |
|
|
|
| C40H66N2O30 | 1054.37 | 4 | 0 | 0 |
3 | M5 |
|
|
|
| C46H76N2O35 | 1216.42 | 5 | 0 | 0 |
4 | M6 |
|
|
|
| C52H86N2O40 | 1378.48 | 6 | 0 | 0 |
5 | M7 |
|
|
|
| C58H96N2O45 | 1540.53 | 7 | 0 | 0 |
6 | M8 |
|
|
|
| C64H106N2O50 | 1702.58 | 8 | 0 | 0 |
7 | M9 |
|
|
|
| C70H116N2O55 | 1864.63 | 9 | 0 | 0 |
8 | A1G0 | Gn- |
|
|
| C42H69N3O30 | 1095.4 | 3 | 0 | 0 |
9 | A1G0F | Gn- |
|
|
| C48H79N3O34 | 1241.45 | 3 | 1 | 0 |
10 | A2G0 | Gn- |
| Gn- |
| C50H82N4O35 | 1298.48 | 3 | 0 | 0 |
11 | A2G0B | Gn- |
| Gn- |
| C58H95N5O40 | 1501.56 | 3 | 0 | 1 |
12 | A2G0F | Gn- |
| Gn- |
| C56H92N4O39 | 1444.53 | 3 | 1 | 0 |
13 | A2G0FB | Gn- |
| Gn- |
| C64H105N5O44 | 1647.61 | 3 | 1 | 1 |
14 | A1G1 | G-Gn- |
|
|
| C48H79N3O35 | 1257.45 | 3 | 0 | 0 |
15 | A1G1F | G-Gn- |
|
|
| C54H89N3O39 | 1403.51 | 3 | 1 | 0 |
16 | A2G1 | G-Gn- |
| Gn- |
| C56H92N4O40 | 1460.53 | 3 | 0 | 0 |
17 | A2G1B | G-Gn- |
| Gn- |
| C64H105N5O45 | 1663.61 | 3 | 0 | 1 |
18 | A2G1F | G-Gn- |
| Gn- |
| C62H102N4O44 | 1606.59 | 3 | 1 | 0 |
19 | A2G1FB | G-Gn- |
| Gn- |
| C70H115N5O49 | 1809.67 | 3 | 1 | 1 |
20 | A2G2 | G-Gn- |
| G-Gn- |
| C62H102N4O45 | 1622.58 | 3 | 0 | 0 |
21 | A2G2B | G-Gn- |
| G-Gn- |
| C70H115N5O50 | 1825.66 | 3 | 0 | 1 |
22 | A2G2F | G-Gn- |
| G-Gn- |
| C68H112N4O49 | 1768.64 | 3 | 1 | 0 |
23 | A2G2FB | G-Gn- |
| G-Gn- |
| C76H125N5O54 | 1971.72 | 3 | 1 | 1 |
24 | A1S1 | S-G-Gn- |
|
|
| C59H96N4O43 | 1548.54 | 3 | 0 | 0 |
25 | A1S1F | S-G-Gn- |
|
|
| C65H106N4O47 | 1694.6 | 3 | 1 | 0 |
26 | A2S1G0 | S-G-Gn- |
| Gn- |
| C67H109N5O48 | 1751.62 | 3 | 0 | 0 |
27 | A2S1G0B | S-G-Gn- |
| Gn- |
| C75H122N6O53 | 1954.7 | 3 | 0 | 1 |
28 | A2S1G0F | S-G-Gn- |
| Gn- |
| C73H119N5O52 | 1897.68 | 3 | 1 | 0 |
29 | A2S1G0FB | S-G-Gn- |
| Gn- |
| C81H132N6O57 | 2100.76 | 3 | 1 | 1 |
30 | A2S1G1 | S-G-Gn- |
| G-Gn- |
| C73H119N5O53 | 1913.68 | 3 | 0 | 0 |
31 | A2S1G1B | S-G-Gn- |
| G-Gn- |
| C81H132N6O58 | 2116.76 | 3 | 0 | 1 |
32 | A2S1G1F | S-G-Gn- |
| G-Gn- |
| C79H129N5O57 | 2059.73 | 3 | 1 | 0 |
33 | A2S1G1FB | S-G-Gn- |
| G-Gn- |
| C87H142N6O62 | 2262.81 | 3 | 1 | 1 |
34 | A2S2 | S-G-Gn- |
| S-G-Gn- |
| C84H136N6O61 | 2204.77 | 3 | 0 | 0 |
35 | A2S2B | S-G-Gn- |
| S-G-Gn- |
| C92H149N7O66 | 2407.85 | 3 | 0 | 1 |
36 | A2S2F | S-G-Gn- |
| S-G-Gn- |
| C90H146N6O65 | 2350.83 | 3 | 1 | 0 |
37 | A2S2FB | S-G-Gn- |
| S-G-Gn- |
| C98H159N7O70 | 2553.91 | 3 | 1 | 1 |
38 | Gn |
|
|
|
| C8H13N1O5 | 203.079 | 0 | 0 | 0 |
39 | GnF |
|
|
|
| C14H23N1O9 | 349.137 | 0 | 1 | 0 |