The following table lists the N-linked glycans, sorted by CHO host cell-line type, that are included in the N-glycan-specific search.
In the table header, "Mono mass" refers to monoisotopic mass, "Man" refers to mannose, "Fuc" refers to fucose, and "Bis Gn" refers to bisecting glycoprotein.
# | Glycan name | Antenna | Formula | Mono mass | Man | Fuc | Bis Gn | |||
|---|---|---|---|---|---|---|---|---|---|---|
1 | 2 | 3 | 4 | |||||||
1 | M3 |
|
|
|
| C34H56N2O25 | 892.317 | 3 | 0 | 0 |
2 | M4 |
|
|
|
| C40H66N2O30 | 1054.37 | 4 | 0 | 0 |
3 | M5 |
|
|
|
| C46H76N2O35 | 1216.42 | 5 | 0 | 0 |
4 | M6 |
|
|
|
| C52H86N2O40 | 1378.48 | 6 | 0 | 0 |
5 | M7 |
|
|
|
| C58H96N2O45 | 1540.53 | 7 | 0 | 0 |
6 | M8 |
|
|
|
| C64H106N2O50 | 1702.58 | 8 | 0 | 0 |
7 | M9 |
|
|
|
| C70H116N2O55 | 1864.63 | 9 | 0 | 0 |
8 | A1G0 | Gn- |
|
|
| C42H69N3O30 | 1095.4 | 3 | 0 | 0 |
9 | A1G0F | Gn- |
|
|
| C48H79N3O34 | 1241.45 | 3 | 1 | 0 |
10 | A1G0M4 | Gn- |
|
|
| C48H79N3O35 | 1257.45 | 4 | 0 | 0 |
11 | A1G0M4F | Gn- |
|
|
| C54H89N3O39 | 1403.51 | 4 | 1 | 0 |
12 | A1G0M5 | Gn- |
|
|
| C54H89N3O40 | 1419.5 | 5 | 0 | 0 |
13 | A1G0M5F | Gn- |
|
|
| C60H99N3O44 | 1565.56 | 5 | 1 | 0 |
14 | A2G0 | Gn- |
| Gn- |
| C50H82N4O35 | 1298.48 | 3 | 0 | 0 |
15 | A2G0F | Gn- |
| Gn- |
| C56H92N4O39 | 1444.53 | 3 | 1 | 0 |
16 | A2G0M4 | Gn- | Gn- |
|
| C56H92N4O40 | 1460.53 | 4 | 0 | 0 |
17 | A2G0M5 | Gn- | Gn- |
|
| C62H102N4O45 | 1622.58 | 5 | 0 | 0 |
18 | A2G0M5F | Gn- | Gn- |
|
| C68H112N4O49 | 1768.64 | 5 | 1 | 0 |
19 | A3G0 | Gn- | Gn- | Gn- |
| C58H95N5O40 | 1501.56 | 3 | 0 | 0 |
20 | A3G0F | Gn- | Gn- | Gn- |
| C64H105N5O44 | 1647.61 | 3 | 1 | 0 |
21 | A4G0 | Gn- | Gn- | Gn- | Gn- | C66H108N6O45 | 1704.63 | 3 | 0 | 0 |
22 | A4G0F | Gn- | Gn- | Gn- | Gn- | C72H118N6O49 | 1850.69 | 3 | 1 | 0 |
23 | A1G1 | G-Gn- |
|
|
| C48H79N3O35 | 1257.45 | 3 | 0 | 0 |
24 | A1G1F | G-Gn- |
|
|
| C54H89N3O39 | 1403.51 | 3 | 1 | 0 |
25 | A1G1M4 | G-Gn- |
|
|
| C54H89N3O40 | 1419.5 | 4 | 0 | 0 |
26 | A1G1M4F | G-Gn- |
|
|
| C60H99N3O44 | 1565.56 | 4 | 1 | 0 |
27 | A1G1M5 | G-Gn- |
|
|
| C60H99N3O45 | 1581.56 | 5 | 0 | 0 |
28 | A1G1M5F | G-Gn- |
|
|
| C66H109N3O49 | 1727.61 | 5 | 1 | 0 |
29 | A2G1 | G-Gn- |
| Gn- |
| C56H92N4O40 | 1460.53 | 3 | 0 | 0 |
30 | A2G1F | G-Gn- |
| Gn- |
| C62H102N4O44 | 1606.59 | 3 | 1 | 0 |
31 | A2G2 | G-Gn- |
| G-Gn- |
| C62H102N4O45 | 1622.58 | 3 | 0 | 0 |
32 | A2G2F | G-Gn- |
| G-Gn- |
| C68H112N4O49 | 1768.64 | 3 | 1 | 0 |
33 | A2G1M4 | G-Gn- | Gn- |
|
| C62H102N4O45 | 1622.58 | 4 | 0 | 0 |
34 | A2G1M4F | G-Gn- | Gn- |
|
| C68H112N4O49 | 1768.64 | 4 | 1 | 0 |
35 | A2G1M5 | G-Gn- | Gn- |
|
| C68H112N4O50 | 1784.63 | 5 | 0 | 0 |
36 | A2G1M5F | G-Gn- | Gn- |
|
| C74H122N4O54 | 1930.69 | 5 | 1 | 0 |
37 | A3G1 | G-Gn- | Gn- | Gn- |
| C64H105N5O45 | 1663.61 | 3 | 0 | 0 |
38 | A3G1F | G-Gn- | Gn- | Gn- |
| C70H115N5O49 | 1809.67 | 3 | 1 | 0 |
39 | A4G1 | G-Gn- | Gn- | Gn- | Gn- | C72H118N6O50 | 1866.69 | 3 | 0 | 0 |
40 | A4G1F | G-Gn- | Gn- | Gn- | Gn- | C78H128N6O54 | 2012.75 | 3 | 1 | 0 |
41 | A2G2M4 | G-Gn- | G-Gn- |
|
| C68H112N4O50 | 1784.63 | 4 | 0 | 0 |
42 | A2G2M4F | G-Gn- | G-Gn- |
|
| C74H122N4O54 | 1930.69 | 4 | 1 | 0 |
43 | A2G2M5 | G-Gn- | G-Gn- |
|
| C74H122N4O55 | 1946.69 | 5 | 0 | 0 |
44 | A2G2M5F | G-Gn- | G-Gn- |
|
| C80H132N4O59 | 2092.75 | 5 | 1 | 0 |
45 | A3G2 | G-Gn- | G-Gn- | Gn- |
| C70H115N5O50 | 1825.66 | 3 | 0 | 0 |
46 | A3G2F | G-Gn- | G-Gn- | Gn- |
| C76H125N5O54 | 1971.72 | 3 | 1 | 0 |
47 | A4G2 | G-Gn- | G-Gn- | Gn- | Gn- | C78H128N6O55 | 2028.74 | 3 | 0 | 0 |
48 | A4G2F | G-Gn- | G-Gn- | Gn- | Gn- | C84H138N6O59 | 2174.8 | 3 | 1 | 0 |
49 | A3G3 | G-Gn- | G-Gn- | G-Gn- |
| C76H125N5O55 | 1987.71 | 3 | 0 | 0 |
50 | A3G3F | G-Gn- | G-Gn- | G-Gn- |
| C82H135N5O59 | 2133.77 | 3 | 1 | 0 |
51 | A4G3 | G-Gn- | G-Gn- | G-Gn- | Gn- | C84H138N6O60 | 2190.79 | 3 | 0 | 0 |
52 | A4G3F | G-Gn- | G-Gn- | G-Gn- | Gn- | C90H148N6O64 | 2336.85 | 3 | 1 | 0 |
53 | A4G4 | G-Gn- | G-Gn- | G-Gn- | G-Gn- | C90H148N6O65 | 2352.85 | 3 | 0 | 0 |
54 | A4G4F | G-Gn- | G-Gn- | G-Gn- | G-Gn- | C96H158N6O69 | 2498.9 | 3 | 1 | 0 |
55 | A1S1 | S-G-Gn- |
|
|
| C59H96N4O43 | 1548.54 | 3 | 0 | 0 |
56 | A1S1F | S-G-Gn- |
|
|
| C65H106N4O47 | 1694.6 | 3 | 1 | 0 |
57 | A1S1M4 | S-G-Gn- |
|
|
| C65H106N4O48 | 1710.6 | 4 | 0 | 0 |
58 | A1S1M4F | S-G-Gn- |
|
|
| C71H116N4O52 | 1856.66 | 4 | 1 | 0 |
59 | A1S1M5 | S-G-Gn- |
|
|
| C71H116N4O53 | 1872.65 | 5 | 0 | 0 |
60 | A1S1M5F | S-G-Gn- |
|
|
| C77H126N4O57 | 2018.71 | 5 | 1 | 0 |
61 | A2S1G0 | S-G-Gn- |
| Gn- |
| C67H109N5O48 | 1751.62 | 3 | 0 | 0 |
62 | A2S1G0F | S-G-Gn- |
| Gn- |
| C73H119N5O52 | 1897.68 | 3 | 1 | 0 |
63 | A2S1G1 | S-G-Gn- |
| G-Gn- |
| C73H119N5O53 | 1913.68 | 3 | 0 | 0 |
64 | A2S1G1F | S-G-Gn- |
| G-Gn- |
| C79H129N5O57 | 2059.73 | 3 | 1 | 0 |
65 | A2S2 | S-G-Gn- |
| S-G-Gn- |
| C84H136N6O61 | 2204.77 | 3 | 0 | 0 |
66 | A2S2F | S-G-Gn- |
| S-G-Gn- |
| C90H146N6O65 | 2350.83 | 3 | 1 | 0 |
67 | A2S1G0M4 | S-G-Gn- | Gn- |
|
| C73H119N5O53 | 1913.68 | 4 | 0 | 0 |
68 | A2S1G0M4F | S-G-Gn- | Gn- |
|
| C79H129N5O57 | 2059.73 | 4 | 1 | 0 |
69 | A2S1G0M5 | S-G-Gn- | Gn- |
|
| C79H129N5O58 | 2075.73 | 5 | 0 | 0 |
70 | A2S1G0M5F | S-G-Gn- | Gn- |
|
| C85H139N5O62 | 2221.79 | 5 | 1 | 0 |
71 | A3S1G0 | S-G-Gn- | Gn- | Gn- |
| C75H122N6O53 | 1954.7 | 3 | 0 | 0 |
72 | A3S1G0F | S-G-Gn- | Gn- | Gn- |
| C81H132N6O57 | 2100.76 | 3 | 1 | 0 |
73 | A4S1G0 | S-G-Gn- | Gn- | Gn- | Gn- | C83H135N7O58 | 2157.78 | 3 | 0 | 0 |
74 | A4S1G0F | S-G-Gn- | Gn- | Gn- | Gn- | C89H145N7O62 | 2303.84 | 3 | 1 | 0 |
75 | A2S1G1M4 | S-G-Gn- | G-Gn- |
|
| C79H129N5O58 | 2075.73 | 4 | 0 | 0 |
76 | A2S1G1M4F | S-G-Gn- | G-Gn- |
|
| C85H139N5O62 | 2221.79 | 4 | 1 | 0 |
77 | A2S1G1M5 | S-G-Gn- | G-Gn- |
|
| C85H139N5O63 | 2237.78 | 5 | 0 | 0 |
78 | A2S1G1M5F | S-G-Gn- | G-Gn- |
|
| C91H149N5O67 | 2383.84 | 5 | 1 | 0 |
79 | A3S1G1 | S-G-Gn- | G-Gn- | Gn- |
| C81H132N6O58 | 2116.76 | 3 | 0 | 0 |
80 | A3S1G1F | S-G-Gn- | G-Gn- | Gn- |
| C87H142N6O62 | 2262.81 | 3 | 1 | 0 |
81 | A4S1G1 | S-G-Gn- | G-Gn- | Gn- | Gn- | C89H145N7O63 | 2319.84 | 3 | 0 | 0 |
82 | A4S1G1F | S-G-Gn- | G-Gn- | Gn- | Gn- | C95H155N7O67 | 2465.89 | 3 | 1 | 0 |
83 | A3S1G2 | S-G-Gn- | G-Gn- | G-Gn- |
| C87H142N6O63 | 2278.81 | 3 | 0 | 0 |
84 | A3S1G2F | S-G-Gn- | G-Gn- | G-Gn- |
| C93H152N6O67 | 2424.87 | 3 | 1 | 0 |
85 | A4S1G2 | S-G-Gn- | G-Gn- | G-Gn- | Gn- | C95H155N7O68 | 2481.89 | 3 | 0 | 0 |
86 | A4S1G2F | S-G-Gn- | G-Gn- | G-Gn- | Gn- | C101H165N7O72 | 2627.95 | 3 | 1 | 0 |
87 | A4S1G3 | S-G-Gn- | G-Gn- | G-Gn- | G-Gn- | C101H165N7O73 | 2643.94 | 3 | 0 | 0 |
88 | A4S1G3F | S-G-Gn- | G-Gn- | G-Gn- | G-Gn- | C107H175N7O77 | 2790 | 3 | 1 | 0 |
89 | A2S2M4 | S-G-Gn- | S-G-Gn- |
|
| C90H146N6O66 | 2366.83 | 4 | 0 | 0 |
90 | A2S2M4F | S-G-Gn- | S-G-Gn- |
|
| C96H156N6O70 | 2512.88 | 4 | 1 | 0 |
91 | A2S2M5 | S-G-Gn- | S-G-Gn- |
|
| C96H156N6O71 | 2528.88 | 5 | 0 | 0 |
92 | A2S2M5F | S-G-Gn- | S-G-Gn- |
|
| C102H166N6O75 | 2674.94 | 5 | 1 | 0 |
93 | A3S2G0 | S-G-Gn- | S-G-Gn- | Gn- |
| C92H149N7O66 | 2407.85 | 3 | 0 | 0 |
94 | A3S2G0F | S-G-Gn- | S-G-Gn- | Gn- |
| C98H159N7O70 | 2553.91 | 3 | 1 | 0 |
95 | A4S2G0 | S-G-Gn- | S-G-Gn- | Gn- | Gn- | C100H162N8O71 | 2610.93 | 3 | 0 | 0 |
96 | A4S2G0F | S-G-Gn- | S-G-Gn- | Gn- | Gn- | C106H172N8O75 | 2756.99 | 3 | 1 | 0 |
97 | A3S2G1 | S-G-Gn- | S-G-Gn- | G-Gn- |
| C98H159N7O71 | 2569.9 | 3 | 0 | 0 |
98 | A3S2G1F | S-G-Gn- | S-G-Gn- | G-Gn- |
| C104H169N7O75 | 2715.96 | 3 | 1 | 0 |
99 | A4S2G1 | S-G-Gn- | S-G-Gn- | G-Gn- | Gn- | C106H172N8O76 | 2772.98 | 3 | 0 | 0 |
100 | A4S2G1F | S-G-Gn- | S-G-Gn- | G-Gn- | Gn- | C112H182N8O80 | 2919.04 | 3 | 1 | 0 |
101 | A4S2G2 | S-G-Gn- | S-G-Gn- | G-Gn- | G-Gn- | C112H182N8O81 | 2935.04 | 3 | 0 | 0 |
102 | A4S2G2F | S-G-Gn- | S-G-Gn- | G-Gn- | G-Gn- | C118H192N8O85 | 3081.09 | 3 | 1 | 0 |
103 | A3S3 | S-G-Gn- | S-G-Gn- | S-G-Gn- |
| C109H176N8O79 | 2861 | 3 | 0 | 0 |
104 | A3S3F | S-G-Gn- | S-G-Gn- | S-G-Gn- |
| C115H186N8O83 | 3007.06 | 3 | 1 | 0 |
105 | A4S3G0 | S-G-Gn- | S-G-Gn- | S-G-Gn- | Gn- | C117H189N9O84 | 3064.08 | 3 | 0 | 0 |
106 | A4S3G0F | S-G-Gn- | S-G-Gn- | S-G-Gn- | Gn- | C123H199N9O88 | 3210.14 | 3 | 1 | 0 |
107 | A4S3G1 | S-G-Gn- | S-G-Gn- | S-G-Gn- | G-Gn- | C123H199N9O89 | 3226.13 | 3 | 0 | 0 |
108 | A4S3G1F | S-G-Gn- | S-G-Gn- | S-G-Gn- | G-Gn- | C129H209N9O93 | 3372.19 | 3 | 1 | 0 |
109 | A4S4 | S-G-Gn- | S-G-Gn- | S-G-Gn- | S-G-Gn- | C134H216N10O97 | 3517.23 | 3 | 0 | 0 |
110 | A4S4F | S-G-Gn- | S-G-Gn- | S-G-Gn- | S-G-Gn- | C140H226N10O101 | 3663.29 | 3 | 1 | 0 |
111 | A1Sg1 | Sg-G-Gn- |
|
|
| C59H96N4O44 | 1564.54 | 3 | 0 | 0 |
112 | A1Sg1F | Sg-G-Gn- |
|
|
| C65H106N4O48 | 1710.6 | 3 | 1 | 0 |
113 | A2Sg1G0 | Sg-G-Gn- |
| Gn- |
| C67H109N5O49 | 1767.62 | 3 | 0 | 0 |
114 | A2Sg1G0F | Sg-G-Gn- |
| Gn- |
| C73H119N5O53 | 1913.68 | 3 | 1 | 0 |
115 | A2Sg1G1 | Sg-G-Gn- |
| G-Gn- |
| C73H119N5O54 | 1929.67 | 3 | 0 | 0 |
116 | A2Sg1G1F | Sg-G-Gn- |
| G-Gn- |
| C79H129N5O58 | 2075.73 | 3 | 1 | 0 |
117 | A2Sg1S1 | Sg-G-Gn- |
| S-G-Gn- |
| C84H136N6O62 | 2220.77 | 3 | 0 | 0 |
118 | A2Sg1S1F | Sg-G-Gn- |
| S-G-Gn- |
| C90H146N6O66 | 2366.83 | 3 | 1 | 0 |
119 | A2Sg2 | Sg-G-Gn- |
| Sg-G-Gn- |
| C84H136N6O63 | 2236.76 | 3 | 0 | 0 |
120 | A2Sg2F | Sg-G-Gn- |
| Sg-G-Gn- |
| C90H146N6O67 | 2382.82 | 3 | 1 | 0 |
121 | A3Sg1G0 | Sg-G-Gn- | Gn- | Gn- |
| C75H122N6O54 | 1970.7 | 3 | 0 | 0 |
122 | A3Sg1G0F | Sg-G-Gn- | Gn- | Gn- |
| C81H132N6O58 | 2116.76 | 3 | 1 | 0 |
123 | A4Sg1G0 | Sg-G-Gn- | Gn- | Gn- | Gn- | C83H135N7O59 | 2173.78 | 3 | 0 | 0 |
124 | A4Sg1G0F | Sg-G-Gn- | Gn- | Gn- | Gn- | C89H145N7O63 | 2319.84 | 3 | 1 | 0 |
125 | A3Sg1G1 | Sg-G-Gn- | G-Gn- | Gn- |
| C81H132N6O59 | 2132.75 | 3 | 0 | 0 |
126 | A3Sg1G1F | Sg-G-Gn- | G-Gn- | Gn- |
| C87H142N6O63 | 2278.81 | 3 | 1 | 0 |
127 | A4Sg1G1 | Sg-G-Gn- | G-Gn- | Gn- | Gn- | C89H145N7O64 | 2335.83 | 3 | 0 | 0 |
128 | A4Sg1G1F | Sg-G-Gn- | G-Gn- | Gn- | Gn- | C95H155N7O68 | 2481.89 | 3 | 1 | 0 |
129 | A3Sg1G2 | Sg-G-Gn- | G-Gn- | G-Gn- |
| C87H142N6O64 | 2294.8 | 3 | 0 | 0 |
130 | A3Sg1G2F | Sg-G-Gn- | G-Gn- | G-Gn- |
| C93H152N6O68 | 2440.86 | 3 | 1 | 0 |
131 | A4Sg1G2 | Sg-G-Gn- | G-Gn- | G-Gn- | Gn- | C95H155N7O69 | 2497.88 | 3 | 0 | 0 |
132 | A4Sg1G2F | Sg-G-Gn- | G-Gn- | G-Gn- | Gn- | C101H165N7O73 | 2643.94 | 3 | 1 | 0 |
133 | A4Sg1G3 | Sg-G-Gn- | G-Gn- | G-Gn- | G-Gn- | C101H165N7O74 | 2659.94 | 3 | 0 | 0 |
134 | A4Sg1G3F | Sg-G-Gn- | G-Gn- | G-Gn- | G-Gn- | C107H175N7O78 | 2805.99 | 3 | 1 | 0 |
135 | A3Sg1S1G0 | Sg-G-Gn- | S-G-Gn- | Gn- |
| C92H149N7O67 | 2423.85 | 3 | 0 | 0 |
136 | A3Sg1S1G0F | Sg-G-Gn- | S-G-Gn- | Gn- |
| C98H159N7O71 | 2569.9 | 3 | 1 | 0 |
137 | A4Sg1S1G0 | Sg-G-Gn- | S-G-Gn- | Gn- | Gn- | C100H162N8O72 | 2626.93 | 3 | 0 | 0 |
138 | A4Sg1S1G0F | Sg-G-Gn- | S-G-Gn- | Gn- | Gn- | C106H172N8O76 | 2772.98 | 3 | 1 | 0 |
139 | A3Sg1S1G1 | Sg-G-Gn- | S-G-Gn- | G-Gn- |
| C98H159N7O72 | 2585.9 | 3 | 0 | 0 |
140 | A3Sg1S1G1F | Sg-G-Gn- | S-G-Gn- | G-Gn- |
| C104H169N7O76 | 2731.96 | 3 | 1 | 0 |
141 | A4Sg1S1G1 | Sg-G-Gn- | S-G-Gn- | G-Gn- | Gn- | C106H172N8O77 | 2788.98 | 3 | 0 | 0 |
142 | A4Sg1S1G1F | Sg-G-Gn- | S-G-Gn- | G-Gn- | Gn- | C112H182N8O81 | 2935.04 | 3 | 1 | 0 |
143 | A4Sg1S1G2 | Sg-G-Gn- | S-G-Gn- | G-Gn- | G-Gn- | C112H182N8O82 | 2951.03 | 3 | 0 | 0 |
144 | A4Sg1S1G2F | Sg-G-Gn- | S-G-Gn- | G-Gn- | G-Gn- | C118H192N8O86 | 3097.09 | 3 | 1 | 0 |
145 | A3Sg1S2 | Sg-G-Gn- | S-G-Gn- | S-G-Gn- |
| C109H176N8O80 | 2877 | 3 | 0 | 0 |
146 | A3Sg1S2F | Sg-G-Gn- | S-G-Gn- | S-G-Gn- |
| C115H186N8O84 | 3023.05 | 3 | 1 | 0 |
147 | A4Sg1S2G0 | Sg-G-Gn- | S-G-Gn- | S-G-Gn- | Gn- | C117H189N9O85 | 3080.07 | 3 | 0 | 0 |
148 | A4Sg1S2G0F | Sg-G-Gn- | S-G-Gn- | S-G-Gn- | Gn- | C123H199N9O89 | 3226.13 | 3 | 1 | 0 |
149 | A4Sg1S2G1 | Sg-G-Gn- | S-G-Gn- | S-G-Gn- | G-Gn- | C123H199N9O90 | 3242.13 | 3 | 0 | 0 |
150 | A4Sg1S2G1F | Sg-G-Gn- | S-G-Gn- | S-G-Gn- | G-Gn- | C129H209N9O94 | 3388.19 | 3 | 1 | 0 |
151 | A4Sg1S3 | Sg-G-Gn- | S-G-Gn- | S-G-Gn- | S-G-Gn- | C134H216N10O98 | 3533.22 | 3 | 0 | 0 |
152 | A4Sg1S3F | Sg-G-Gn- | S-G-Gn- | S-G-Gn- | S-G-Gn- | C140H226N10O102 | 3679.28 | 3 | 1 | 0 |
153 | A3Sg2G0 | Sg-G-Gn- | Sg-G-Gn- | Gn- |
| C92H149N7O68 | 2439.84 | 3 | 0 | 0 |
154 | A3Sg2G0F | Sg-G-Gn- | Sg-G-Gn- | Gn- |
| C98H159N7O72 | 2585.9 | 3 | 1 | 0 |
155 | A4Sg2G0 | Sg-G-Gn- | Sg-G-Gn- | Gn- | Gn- | C100H162N8O73 | 2642.92 | 3 | 0 | 0 |
156 | A4Sg2G0F | Sg-G-Gn- | Sg-G-Gn- | Gn- | Gn- | C106H172N8O77 | 2788.98 | 3 | 1 | 0 |
157 | A3Sg2G1 | Sg-G-Gn- | Sg-G-Gn- | G-Gn- |
| C98H159N7O73 | 2601.89 | 3 | 0 | 0 |
158 | A3Sg2G1F | Sg-G-Gn- | Sg-G-Gn- | G-Gn- |
| C104H169N7O77 | 2747.95 | 3 | 1 | 0 |
159 | A4Sg2G1 | Sg-G-Gn- | Sg-G-Gn- | G-Gn- | Gn- | C106H172N8O78 | 2804.97 | 3 | 0 | 0 |
160 | A4Sg2G1F | Sg-G-Gn- | Sg-G-Gn- | G-Gn- | Gn- | C112H182N8O82 | 2951.03 | 3 | 1 | 0 |
161 | A4Sg2G2 | Sg-G-Gn- | Sg-G-Gn- | G-Gn- | G-Gn- | C112H182N8O83 | 2967.03 | 3 | 0 | 0 |
162 | A4Sg2G2F | Sg-G-Gn- | Sg-G-Gn- | G-Gn- | G-Gn- | C118H192N8O87 | 3113.08 | 3 | 1 | 0 |
163 | A3Sg2S1 | Sg-G-Gn- | Sg-G-Gn- | S-G-Gn- |
| C109H176N8O81 | 2892.99 | 3 | 0 | 0 |
164 | A3Sg2S1F | Sg-G-Gn- | Sg-G-Gn- | S-G-Gn- |
| C115H186N8O85 | 3039.05 | 3 | 1 | 0 |
165 | A4Sg2S1G0 | Sg-G-Gn- | Sg-G-Gn- | S-G-Gn- | Gn- | C117H189N9O86 | 3096.07 | 3 | 0 | 0 |
166 | A4Sg2S1G0F | Sg-G-Gn- | Sg-G-Gn- | S-G-Gn- | Gn- | C123H199N9O90 | 3242.13 | 3 | 1 | 0 |
167 | A4Sg2S1G1 | Sg-G-Gn- | Sg-G-Gn- | S-G-Gn- | G-Gn- | C123H199N9O91 | 3258.12 | 3 | 0 | 0 |
168 | A4Sg2S1G1F | Sg-G-Gn- | Sg-G-Gn- | S-G-Gn- | G-Gn- | C129H209N9O95 | 3404.18 | 3 | 1 | 0 |
169 | A4Sg2S2 | Sg-G-Gn- | Sg-G-Gn- | S-G-Gn- | S-G-Gn- | C134H216N10O99 | 3549.22 | 3 | 0 | 0 |
170 | A4Sg2S2F | Sg-G-Gn- | Sg-G-Gn- | S-G-Gn- | S-G-Gn- | C140H226N10O103 | 3695.28 | 3 | 1 | 0 |
171 | A3Sg3 | Sg-G-Gn- | Sg-G-Gn- | Sg-G-Gn- |
| C109H176N8O82 | 2908.98 | 3 | 0 | 0 |
172 | A3Sg3F | Sg-G-Gn- | Sg-G-Gn- | Sg-G-Gn- |
| C115H186N8O86 | 3055.04 | 3 | 1 | 0 |
173 | A4Sg3G0 | Sg-G-Gn- | Sg-G-Gn- | Sg-G-Gn- | Gn- | C117H189N9O87 | 3112.06 | 3 | 0 | 0 |
174 | A4Sg3G0F | Sg-G-Gn- | Sg-G-Gn- | Sg-G-Gn- | Gn- | C123H199N9O91 | 3258.12 | 3 | 1 | 0 |
175 | A4Sg3G1 | Sg-G-Gn- | Sg-G-Gn- | Sg-G-Gn- | G-Gn- | C123H199N9O92 | 3274.12 | 3 | 0 | 0 |
176 | A4Sg3G1F | Sg-G-Gn- | Sg-G-Gn- | Sg-G-Gn- | G-Gn- | C129H209N9O96 | 3420.18 | 3 | 1 | 0 |
177 | A4Sg3S1 | Sg-G-Gn- | Sg-G-Gn- | Sg-G-Gn- | S-G-Gn- | C134H216N10O100 | 3565.21 | 3 | 0 | 0 |
178 | A4Sg3S1F | Sg-G-Gn- | Sg-G-Gn- | Sg-G-Gn- | S-G-Gn- | C140H226N10O104 | 3711.27 | 3 | 1 | 0 |
179 | A4Sg4 | Sg-G-Gn- | Sg-G-Gn- | Sg-G-Gn- | Sg-G-Gn- | C134H216N10O101 | 3581.21 | 3 | 0 | 0 |
180 | A4Sg4F | Sg-G-Gn- | Sg-G-Gn- | Sg-G-Gn- | Sg-G-Gn- | C140H226N10O105 | 3727.27 | 3 | 1 | 0 |
181 | Gn |
|
|
|
| C8H13N1O5 | 203.079 | 0 | 0 | 0 |
182 | GnF |
|
|
|
| C14H23N1O9 | 349.137 | 0 | 1 | 0 |